Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3α-Hydroxymogrol | 1343402-73-2 | 5 MG
SDP

Catalog No. 5000418347
Encompass_Preferred

3α-Hydroxymogrol is a triterpenoid isolated from *Siraitia grosvenorii* Swingle. It acts as a potent AMPK activator, enhancing AMPK phosphorylation. This product is for research use only and not sold to patients.

  • Potent AMPK activator
  • Enhances AMPK phosphorylation
  • Triterpenoid structure
  • Isolated from *Siraitia grosvenorii* Swingle
  • Molecular weight: 476.73
  • Chemical formula: C30H52O4
  • Appearance: White to off-white solid
  • Target: AMPK

Catalog No. 50-004-18347 Supplier Medchemexpress LLC Supplier No. HYN69135MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3α-Hydroxymogrol is a triterpenoid isolated from *Siraitia grosvenorii* Swingle. It acts as a potent AMPK activator, enhancing AMPK phosphorylation. This product is for research use only and not sold to patients.

  • Potent AMPK activator
  • Enhances AMPK phosphorylation
  • Triterpenoid structure
  • Isolated from *Siraitia grosvenorii* Swingle
  • Molecular weight: 476.73
  • Chemical formula: C30H52O4
  • Appearance: White to off-white solid
  • Target: AMPK

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.