Learn More
Medchemexpress LLC 3-hydroxyoctanoic acid | 14292-27-4 | MFCD00133277 | >98.0% | 160.21 g/mol | C8H16O3 | 5 MG
SDP
Catalog No. 5000264283
Encompass_Preferred
Analytical standard of 3-hydroxyoctanoic acid, a hydroxylated C8 fatty acid used as a reference compound in chromatographic and mass-spectrometry assays. Suitable for qualitative and quantitative HPLC, GC, and MS method development, calibration, and validation.
  • Used as a reference standard in HPLC, GC, and MS.
  • Hydroxylated C8 fatty acid; molecular weight 160.21 g/mol; formula C8H16O3.
  • Reported to act as an agonist of GPR109B and to influence lipolysis in adipocytes.
  • Available in multiple pack sizes; confirm specific quantity before ordering.
  • Store according to the certificate of analysis; shipping typically at room temperature.
Catalog No. 50-002-64283 Supplier Medchemexpress LLC Supplier No. HY1130585MG
Analytical standard of 3-hydroxyoctanoic acid, a hydroxylated C8 fatty acid used as a reference compound in chromatographic and mass-spectrometry assays. Suitable for qualitative and quantitative HPLC, GC, and MS method development, calibration, and validation.
  • Used as a reference standard in HPLC, GC, and MS.
  • Hydroxylated C8 fatty acid; molecular weight 160.21 g/mol; formula C8H16O3.
  • Reported to act as an agonist of GPR109B and to influence lipolysis in adipocytes.
  • Available in multiple pack sizes; confirm specific quantity before ordering.
  • Store according to the certificate of analysis; shipping typically at room temperature.