Learn More
TARGETMOL CHEMICALS INC 3-HYDROXYOCTANOIC ACID 25MG
SDP
Catalog No. 502576196
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 3-Hydroxyoctanoic Acid is one of the hydroxy-carboxylic acids (HCAs) that serve as intermediates of energy metabolism. It is an agonist of the orphan receptor GPR109B. purity: 97%
Catalog No. 50-257-6196 Supplier TARGETMOL CHEMICALS INC Supplier No. T971925MG
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 3-Hydroxyoctanoic Acid is one of the hydroxy-carboxylic acids (HCAs) that serve as intermediates of energy metabolism. It is an agonist of the orphan receptor GPR109B. purity: 97%