Learn More
Selleck Chemical LLC 3-Hydroxyphenyl
SDP
Catalog No. 5000003851
Encompass_Preferred
3-(3-Hydroxyphenyl)propionic acid (m-Hydroxyphenylpropionic acid 3-Hydroxyhydrocinnamic acid) is one of the major metabolites of ingested caffeic acid and of the phenolic degradation products of proanthocyanidins by the microflora in the colon
Catalog No. 50-000-03851 Supplier Selleck Chemical LLC Supplier No. S607425MG
3-(3-Hydroxyphenyl)propionic acid (m-Hydroxyphenylpropionic acid 3-Hydroxyhydrocinnamic acid) is one of the major metabolites of ingested caffeic acid and of the phenolic degradation products of proanthocyanidins by the microflora in the colon