Learn More
Medchemexpress LLC (3-Hydroxyphenyl)boronic acid | 87199-18-6 | 137.93 | 10 G
SDP
Catalog No. 5000449000
Encompass_Preferred
(3-Hydroxyphenyl)boronic acid is a drug intermediate used for the synthesis of 17β-hydroxysteroid dehydrogenase type 1 inhibitors. This compound is intended for research use only.
  • Used in the synthesis of 17β-hydroxysteroid dehydrogenase type 1 inhibitors
  • Light yellow to yellow solid appearance
  • Purity of 99.90%
  • Suitable for both in vitro and in vivo studies with various solubility options
Catalog No. 50-004-49000 Supplier Medchemexpress LLC Supplier No. HYW00273410G
(3-Hydroxyphenyl)boronic acid is a drug intermediate used for the synthesis of 17β-hydroxysteroid dehydrogenase type 1 inhibitors. This compound is intended for research use only.
  • Used in the synthesis of 17β-hydroxysteroid dehydrogenase type 1 inhibitors
  • Light yellow to yellow solid appearance
  • Purity of 99.90%
  • Suitable for both in vitro and in vivo studies with various solubility options