Learn More
Medchemexpress LLC (3-Hydroxyphenyl)boronic acid | 87199-18-6 | 137.93 | 10MM/1ML
SDP
Catalog No. 5000447443
Encompass_Preferred
(3-Hydroxyphenyl)boronic acid is a drug intermediate primarily utilized in the synthesis of 17β-hydroxysteroid dehydrogenase type 1 inhibitors. This compound is intended for research purposes only.
  • Drug intermediate for synthesizing 17β-hydroxysteroid dehydrogenase type 1 inhibitors
  • Solid appearance
  • Light yellow to yellow color
  • Soluble in DMSO (100 mg/mL, 725.01 mM)
  • For research use only
Catalog No. 50-004-47443 Supplier Medchemexpress LLC Supplier No. HYW00273410MM/1ML
(3-Hydroxyphenyl)boronic acid is a drug intermediate primarily utilized in the synthesis of 17β-hydroxysteroid dehydrogenase type 1 inhibitors. This compound is intended for research purposes only.
  • Drug intermediate for synthesizing 17β-hydroxysteroid dehydrogenase type 1 inhibitors
  • Solid appearance
  • Light yellow to yellow color
  • Soluble in DMSO (100 mg/mL, 725.01 mM)
  • For research use only