Learn More
Medchemexpress LLC (3-Hydroxyphenyl)boronic acid | 87199-18-6 | 137.93 | 5 G
SDP
Catalog No. 5000447395
Encompass_Preferred
(3-Hydroxyphenyl)boronic acid, also known as 3-Hydroxybenzeneboronic acid, is a drug intermediate primarily used for the synthesis of 17β-hydroxysteroid dehydrogenase type 1 inhibitors. This compound is for research use only.
  • Used as a drug intermediate
  • Synthesizes 17β-hydroxysteroid dehydrogenase type 1 inhibitors
  • Applicable in Suzuki cross-coupling
  • High purity (99.9%)
  • Relevant for estrogen-dependent disease research
Catalog No. 50-004-47395 Supplier Medchemexpress LLC Supplier No. HYW0027345G
(3-Hydroxyphenyl)boronic acid, also known as 3-Hydroxybenzeneboronic acid, is a drug intermediate primarily used for the synthesis of 17β-hydroxysteroid dehydrogenase type 1 inhibitors. This compound is for research use only.
  • Used as a drug intermediate
  • Synthesizes 17β-hydroxysteroid dehydrogenase type 1 inhibitors
  • Applicable in Suzuki cross-coupling
  • High purity (99.9%)
  • Relevant for estrogen-dependent disease research