Learn More
Apexbio Technology LLC 3-Hydroxyphenylacetic acid 621-37-4 5g
SDP
Catalog No. 502240684
Encompass_Preferred
3-Hydroxyphenylacetic acid (CAS 621-37-4) is an endogenous aromatic organic acid and a secondary intermediate in the metabolic pathways of tyrosine and phenylalanine Evidence suggests that it participates in modulating interactions between the gut microbiota and host metabolic processes and may impact neurotransmitter homeostasis Due to its involvement in these biochemical pathways 3-hydroxyphenylacetic acid is frequently employed as a metabolic biomarker to assess gut microbial activity and to investigate the pathophysiological mechanisms underlying neuro-metabolic disorders
This item is not returnable. View return policy
Catalog No. 50-224-0684 Supplier Apexbio Technology LLC Supplier No. B78675000
3-Hydroxyphenylacetic acid (CAS 621-37-4) is an endogenous aromatic organic acid and a secondary intermediate in the metabolic pathways of tyrosine and phenylalanine Evidence suggests that it participates in modulating interactions between the gut microbiota and host metabolic processes and may impact neurotransmitter homeostasis Due to its involvement in these biochemical pathways 3-hydroxyphenylacetic acid is frequently employed as a metabolic biomarker to assess gut microbial activity and to investigate the pathophysiological mechanisms underlying neuro-metabolic disorders