Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypicolinamide 98+%, Thermo Scientific™

Catalog No. AC166941000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC166941000 100 g
AC166940250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166941000 Supplier Thermo Scientific Chemicals Supplier No. 166941000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 933-90-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00006302
InChI Key VIHUZJYFQOEUMI-UHFFFAOYSA-N
PubChem CID 70277
IUPAC Name 3-hydroxypyridine-2-carboxamide
SMILES NC(=O)C1=NC=CC=C1O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 192°C to 196°C
Color Beige-Gray to Brown
Infrared Spectrum Authentic
Quantity 100 g
Synonym 3-hydroxypicolinamide, 3-hydroxy-2-pyridinecarboxamide, 2-pyridinecarboxamide, 3-hydroxy, 3-hydroxy-pyridine-2-carboxylic acid amide, 3-hydroxy-picolinamide, acmc-209rlh, 3-hydroxy picolinic amide, 2-pyridinecarboxamide,3-hydroxy, 3-hydroxy-2-pyridinecarboxylic acid amide
Formula Weight 138.13
Percent Purity ≥98.0%
Physical Form Powder
Chemical Name or Material 3-Hydroxypicolinamide, 98+%
Show More Show Less
EINECSNumber 213-274-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.