Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypicolinamide 98.0+%, TCI America™
SDP

Catalog No. H07865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07865G Supplier TCI America Supplier No. H07865G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 933-90-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00006302
InChI Key VIHUZJYFQOEUMI-UHFFFAOYSA-N
Synonym 3-hydroxypicolinamide, 3-hydroxy-2-pyridinecarboxamide, 2-pyridinecarboxamide, 3-hydroxy, 3-hydroxy-pyridine-2-carboxylic acid amide, 3-hydroxy-picolinamide, acmc-209rlh, 3-hydroxy picolinic amide, 2-pyridinecarboxamide,3-hydroxy, 3-hydroxy-2-pyridinecarboxylic acid amide
PubChem CID 70277
IUPAC Name 3-hydroxypyridine-2-carboxamide
SMILES NC(=O)C1=NC=CC=C1O

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 5 g
Formula Weight 138.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxypicolinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.