Learn More
Cayman Chemical 3-HydroxypIcolInIc AcId 25g
SDP
Catalog No. 502950496
Encompass_Preferred
A matrix substance for MALDI-MS; has been used in the analysis of oligonucleotides and proteins; has been used as a complexation agent for metal complexes; is an intermediate in the biosynthesis of virginiamycin S1
Catalog No. 50-295-0496 Supplier Cayman Chemical Supplier No. 3453025 G
A matrix substance for MALDI-MS; has been used in the analysis of oligonucleotides and proteins; has been used as a complexation agent for metal complexes; is an intermediate in the biosynthesis of virginiamycin S1