Learn More
Apexbio Technology LLC 3-Hydroxypicolinic acid 874-24-8 500mg
SDP
Catalog No. 5000094591
Encompass_Preferred
3-Hydroxypicolinic acid (CAS 874-24-8) is a pyridine-based small molecule characterized by its capacity to chelate metal ions particularly copper(II) This ligand is commonly employed in the synthesis of metal coordination complexes which have been investigated for their ability to modulate biological activity through interactions involving metal ions and biomolecules Studies on such complexes indicate potential anti-tuberculosis properties attributed to their influence on metal-mediated biochemical pathways 3-Hydroxypicolinic acid is thus of interest in the fields of coordination chemistry and drug discovery supporting research into the bioactivity and molecular mechanisms of metal-based therapeutics
Catalog No. 50-000-94591 Supplier Apexbio Technology LLC Supplier No. C7039500
3-Hydroxypicolinic acid (CAS 874-24-8) is a pyridine-based small molecule characterized by its capacity to chelate metal ions particularly copper(II) This ligand is commonly employed in the synthesis of metal coordination complexes which have been investigated for their ability to modulate biological activity through interactions involving metal ions and biomolecules Studies on such complexes indicate potential anti-tuberculosis properties attributed to their influence on metal-mediated biochemical pathways 3-Hydroxypicolinic acid is thus of interest in the fields of coordination chemistry and drug discovery supporting research into the bioactivity and molecular mechanisms of metal-based therapeutics