Learn More
Medchemexpress LLC 3-hydroxypicolinic acid | 874-24-8 | MFCD00006294 | 99.9% | 139.11 g/mol | C6H5NO3 | 25g
SDP
Catalog No. 5000268814
Encompass_Preferred
3-Hydroxypicolinic acid is a matrix 3-Hydroxypicolinic acid coordinates with metal ions such as copper to form complexes with cytotoxic and antimycobacterial activity 3-Hydroxypicolinic acid is used for the analysis of oligodeoxynucleotides in matrix-assisted laser desorption ionization time-of-flight mass spectrometry (MALDI-TOF MS) 3-Hydroxypicolinic acid is useful for nucleic acid analysis and anti-tuberculosis compound development[1][2][3]
Catalog No. 50-002-68814 Supplier Medchemexpress LLC Supplier No. HYY003025G
3-Hydroxypicolinic acid is a matrix 3-Hydroxypicolinic acid coordinates with metal ions such as copper to form complexes with cytotoxic and antimycobacterial activity 3-Hydroxypicolinic acid is used for the analysis of oligodeoxynucleotides in matrix-assisted laser desorption ionization time-of-flight mass spectrometry (MALDI-TOF MS) 3-Hydroxypicolinic acid is useful for nucleic acid analysis and anti-tuberculosis compound development[1][2][3]