Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypicolinic acid matrix substance for MALDI-MS, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015722 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5722 1 g
11-101-5721 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5722 Supplier MilliporeSigma Supelco Supplier No. 561971G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 3-Hydroxypicolinic Acid
CAS 874-24-8
Linear Formula C6H5 NO3
Technique Mass Spectrometry
MDL Number MFCD00006294
Melting Point 213°C to 218°C
Synonym 3-HPA; 3-Hydroxypyridine-2-carboxylic acid
Type Organic Standard
Solvent or Matrix Neat
Molecular Weight (g/mol) 139.11
Quantity 1 g
Formula Weight 139.11
Percent Purity ≥99% (HPLC)
Grade Matrix Substance for MALDI-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.