Learn More
Medchemexpress LLC HY-N6002 5mg Medchemexpress, 3'-Hydroxypterostilbene CAS:475231-21-1 Purity:>98%
SDP
Catalog No. 501970443
Encompass_Preferred
Medchemexpress, HY-N6002 5mg 3'-Hydroxypterostilbene CAS:475231-21-1 3'-Hydroxypterostilbene, a natural pterostilbene analogue, effectively inhibits the growth of human colon cancer cells (IC50s of 9.0, 40.2, and 70.9 µM for COLO 205, HCT-116, and HT-29 cells, respectively) by inducing apoptosis and autophagy. 3'-Hydroxypterostilbene inhibits the PI3K/Akt/mTOR/p70S6K, and p38MAPK pathways and activates the ERK1/2, JNK1/2 MAPK pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0443 Supplier Medchemexpress LLC Supplier No. HYN60025MG
Medchemexpress, HY-N6002 5mg 3'-Hydroxypterostilbene CAS:475231-21-1 3'-Hydroxypterostilbene, a natural pterostilbene analogue, effectively inhibits the growth of human colon cancer cells (IC50s of 9.0, 40.2, and 70.9 µM for COLO 205, HCT-116, and HT-29 cells, respectively) by inducing apoptosis and autophagy. 3'-Hydroxypterostilbene inhibits the PI3K/Akt/mTOR/p70S6K, and p38MAPK pathways and activates the ERK1/2, JNK1/2 MAPK pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.