Learn More
Medchemexpress LLC 3'-Hydroxypuerarin | 117060-54-5 | 432.38 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN198020MG
Encompass_Preferred
3'-Hydroxypuerarin is an isoflavone isolated from the roots of Pueraria lobata (Willd.) Ohwi. It acts as an antioxidant, exhibiting marked ONOO(-), NO•, and total ROS scavenging activities.
  • Isolated from Pueraria lobata (Willd.) Ohwi.
  • Acts as an antioxidant.
  • Exhibits marked ONOO(-) scavenging activity.
  • Exhibits marked NO• scavenging activity.
  • Exhibits marked total ROS scavenging activities.
Catalog No. 50-004-14760