Learn More
Medchemexpress LLC HY-N1980 5mg Medchemexpress, 3'-Hydroxypuerarin CAS:117060-54-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN19805MG
Encompass_Preferred
Medchemexpress, HY-N1980 5mg 3'-Hydroxypuerarin CAS:117060-54-5 3'-Hydroxypuerarin is an isoflavone isolated from the roots of Pueraria lobata (Willd.) Ohwi. 3'-Hydroxypuerarin is a antioxidant, which shows marked ONOO(-), NO•, total ROS scavenging activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2856