Learn More
Medchemexpress LLC 3-Hydroxystearic aci 25mg | 17773-30-7 | 300.48 g/mol | C18H36O3 | 25 MG
SDP
Catalog No. 5000229498
Encompass_Preferred
3-Hydroxystearic acid (Octadecanoic acid, 3-hydroxy-; CAS 17773-30-7) is a 3-hydroxy derivative of stearic acid supplied for research use. It is a long-chain saturated hydroxy fatty acid provided as a powder and characterized by molecular formula C18H36O3, molecular weight 300.48 g/mol, and HPLC purity 98.25%.
  • Long-chain 3-hydroxy fatty acid for analytical and research applications.
  • High purity: 98.25% (HPLC).
  • Molecular formula C18H36O3; molecular weight 300.48 g/mol.
  • Solid powder form with defined storage conditions for stability.
  • Suitable as an analytical standard or biochemical reagent.
Catalog No. 50-002-29498 Supplier Medchemexpress LLC Supplier No. HY11613325MG
Add to Cart
Add to Cart
3-Hydroxystearic acid (Octadecanoic acid, 3-hydroxy-; CAS 17773-30-7) is a 3-hydroxy derivative of stearic acid supplied for research use. It is a long-chain saturated hydroxy fatty acid provided as a powder and characterized by molecular formula C18H36O3, molecular weight 300.48 g/mol, and HPLC purity 98.25%.
  • Long-chain 3-hydroxy fatty acid for analytical and research applications.
  • High purity: 98.25% (HPLC).
  • Molecular formula C18H36O3; molecular weight 300.48 g/mol.
  • Solid powder form with defined storage conditions for stability.
  • Suitable as an analytical standard or biochemical reagent.