Learn More
Medchemexpress LLC 3α-Hydroxysteroid Dehydrogenase, Microorganism (3α-HSD) | 9028-56-2 | ≥50 U/mg protein | About 28 kDa (SDS-PAGE detection) | 1 KU
SDP
Catalog No. 5000398615
Encompass_Preferred
3α-Hydroxysteroid Dehydrogenase, Microorganism (3α-HSD) is an enzyme that catalyzes the conversion of 3-ketosteroids into 3α-hydroxy compounds. Significant in androgen DHT inactivation, it converts DHT into 3α-androstanediol. This genetically recombinant product, with no animal origin, is used clinically to catalyze hydroxysteroid dehydrogenation and for total bile acid detection.
  • Catalyzes the conversion of 3-ketosteroids into 3α-hydroxy compounds.
  • Inactivates androgen DHT.
  • Genetically recombinant with no animal origin.
  • Used for clinical detection of total bile acid.
  • Optimum pH is 7.0-9.0.
  • Inhibited by Hg2+ and Ag+.
  • Specific activity ≥50 U/mg protein.
Catalog No. 50-003-98615 Supplier Medchemexpress LLC Supplier No. HYP29011KU
3α-Hydroxysteroid Dehydrogenase, Microorganism (3α-HSD) is an enzyme that catalyzes the conversion of 3-ketosteroids into 3α-hydroxy compounds. Significant in androgen DHT inactivation, it converts DHT into 3α-androstanediol. This genetically recombinant product, with no animal origin, is used clinically to catalyze hydroxysteroid dehydrogenation and for total bile acid detection.
  • Catalyzes the conversion of 3-ketosteroids into 3α-hydroxy compounds.
  • Inactivates androgen DHT.
  • Genetically recombinant with no animal origin.
  • Used for clinical detection of total bile acid.
  • Optimum pH is 7.0-9.0.
  • Inhibited by Hg2+ and Ag+.
  • Specific activity ≥50 U/mg protein.