Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-3-Hydroxytetrahydrofuran

Catalog No. 3097531G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-975-31G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 30-975-31G Supplier Sigma Aldrich Supplier No. 3097531G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-3-Hydroxytetrahydrofuran may be used as building block in the synthesis of potent HIV protease inhibitors.

Specifications

CAS 95500-19-9
Melting Point 245°C to 251°C
Molecular Formula C9H10N2O4 -+ H2O
Linear Formula O2NC6H4CH2CH(NH2)CO2H -+ H2O
Refractive Index n20/D 1.45
MDL Number MFCD00150543
Quantity 5 g
Synonym (R)-(-)-Tetrahydro-3-furanol
Molecular Weight (g/mol) 228.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.