Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxytetrahydrofuran, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
453-20-3
C4H8O2
88.11
MFCD00005374
XDPCNPCKDGQBAN-UHFFFAOYSA-N
3-hydroxytetrahydrofuran,tetrahydrofuran-3-ol,3-furanol, tetrahydro,tetrahydro-3-furanol,3-hydroxyoxolane,tetrahydro-furan-3-ol,3-hydroxytetrahydrofurane,3-hydroxy tetrahydrofuran,s-3-hydroxytetrahydrofuran,3-tetrahydrofuranol
9960
oxolan-3-ol
C1COCC1O
453-20-3
C4H8O2
88.11
MFCD00005374
XDPCNPCKDGQBAN-UHFFFAOYSA-N
3-hydroxytetrahydrofuran,tetrahydrofuran-3-ol,3-furanol, tetrahydro,tetrahydro-3-furanol,3-hydroxyoxolane,tetrahydro-furan-3-ol,3-hydroxytetrahydrofurane,3-hydroxy tetrahydrofuran,s-3-hydroxytetrahydrofuran,3-tetrahydrofuranol
9960
oxolan-3-ol
C1COCC1O
Flash Point 81°C
Boiling Point 181°C
Specific Gravity 1.09
Density 1.0900g/mL
Refractive Index 1.4490 to 1.451
Quantity 100g
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 17, 107
Formula Weight 88.11
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-Hydroxytetrahydrofuran
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.