Learn More
Medchemexpress LLC HY-113004 50mg Medchemexpress, 3-Hydroxyvaleric acid CAS:10237-77-1 Purity:>98%
SDP
Catalog No. 502080829
Encompass_Preferred
Medchemexpress, HY-113004 50mg 3-Hydroxyvaleric acid CAS:10237-77-1 3-Hydroxyvaleric acid is a 5-carbon ketone body. 3-Hydroxyvaleric acid is anaplerotic, meaning it can refill the pool of TCA cycle intermediates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0829 Supplier Medchemexpress LLC Supplier No. HY11300450MG
Medchemexpress, HY-113004 50mg 3-Hydroxyvaleric acid CAS:10237-77-1 3-Hydroxyvaleric acid is a 5-carbon ketone body. 3-Hydroxyvaleric acid is anaplerotic, meaning it can refill the pool of TCA cycle intermediates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.