Learn More
Medchemexpress LLC 3-hydroxyvalproic acid | 58888-84-9 | MFCD00869521 | 160.21 g·mol⁻¹ | C8H16O3 | 5 MG
SDP
Catalog No. 5000281053
Encompass_Preferred
3-Hydroxyvalproic acid is a metabolite of valproic acid used as a research reagent to investigate valproic acid metabolism and mitochondrial β-oxidation pathway-related enzymes. It is intended for biochemical and metabolic-disease research where study of drug metabolites and enzyme inhibition is required.
  • Metabolite of valproic acid relevant to metabolism studies.
  • Weak inhibitor of mitochondrial β-oxidation pathway-related enzymes.
  • Suitable for analytical assays and biochemical experiments.
  • Supports metabolic disease and drug metabolism research applications.
  • Chemical identifiers: CAS 58888-84-9; formula C8H16O3; MW 160.21 g·mol⁻¹.
Catalog No. 50-002-81053 Supplier Medchemexpress LLC Supplier No. HYW4004215MG
3-Hydroxyvalproic acid is a metabolite of valproic acid used as a research reagent to investigate valproic acid metabolism and mitochondrial β-oxidation pathway-related enzymes. It is intended for biochemical and metabolic-disease research where study of drug metabolites and enzyme inhibition is required.
  • Metabolite of valproic acid relevant to metabolism studies.
  • Weak inhibitor of mitochondrial β-oxidation pathway-related enzymes.
  • Suitable for analytical assays and biochemical experiments.
  • Supports metabolic disease and drug metabolism research applications.
  • Chemical identifiers: CAS 58888-84-9; formula C8H16O3; MW 160.21 g·mol⁻¹.