Learn More
OHAUS 3 in. Platform with Rubber Cover
Catalog No. 01804427 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-427 Vortex Attachment
1 options
Catalog No. 01-804-427 Supplier OHAUS Supplier No. 83041423
Add to Cart
Add to Cart
Plastic 3 in. (76 mm) platform with rubber cover for mixing and vortexing in beakers, flasks and multiple tubes.
  • The plastic 3 in. platform with rubber cover may be used on all Vortex Genie mixers and Disruptor Genies.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 1, Vortex-Genie Pulse and the Vortex-Genie 2