Learn More
Andwin Scientific 3 INCH PLATFORM RUBBER COVER
SDP
Catalog No. NC0325965
Encompass_Preferred
This plastic platform with rubber cover is ideal for mixing and vortexing in laboratory glasswares including beakers, flasks, and multiple tubes. This product is also compatible with Vortex-Genie 1, Vortex-Genie Pulse, and the Vortex-Genie 2 family of mixers. It measures 3 inches or 76 millimeters.
Catalog No. NC0325965 Supplier Andwin Scientific Supplier No. OK0500902
This plastic platform with rubber cover is ideal for mixing and vortexing in laboratory glasswares including beakers, flasks, and multiple tubes. This product is also compatible with Vortex-Genie 1, Vortex-Genie Pulse, and the Vortex-Genie 2 family of mixers. It measures 3 inches or 76 millimeters.