Learn More
Medchemexpress LLC HY-18569 5g Medchemexpress, 3-Indoleacetic acid Indole-3-acetic acid;3-IAA CAS:87-51-4 Purity:98%
SDP
Catalog No. 501638763
Encompass_Preferred
Medchemexpress,HY-18569 5g 3-Indoleacetic acid Indole-3-acetic acid;3-IAA CAS:87-51-4 Formula:C10H9NO2 Indole-3-acetic acid (3-Indoleacetic acid; IAA) is the most common natural plant growth hormone of the auxin class. It can be added to cell culture medium to induce plant cell elongation and division. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8763 Supplier Medchemexpress LLC Supplier No. HY185695G
Medchemexpress,HY-18569 5g 3-Indoleacetic acid Indole-3-acetic acid;3-IAA CAS:87-51-4 Formula:C10H9NO2 Indole-3-acetic acid (3-Indoleacetic acid; IAA) is the most common natural plant growth hormone of the auxin class. It can be added to cell culture medium to induce plant cell elongation and division. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.