Learn More
Medchemexpress LLC 3-Indoleacetic acid-13C6 | 100849-36-3 | >99.9% | 181.14 | 1 MG
SDP
Catalog No. 5000387772
Encompass_Preferred
3-Indoleacetic acid-13C6 is the 13C labeled version of 3-Indoleacetic acid, a prevalent natural plant growth hormone belonging to the auxin class. It is utilized in various biological applications.
  • Acts as a natural plant growth hormone.
  • Induces plant cell elongation and division in cell culture mediums.
  • Functions as a tracer in research.
  • Serves as an internal standard for quantitative analysis.
  • Compatible with analytical techniques such as NMR, GC-MS, and LC-MS.
Catalog No. 50-003-87772 Supplier Medchemexpress LLC Supplier No. HY18569S41MG
3-Indoleacetic acid-13C6 is the 13C labeled version of 3-Indoleacetic acid, a prevalent natural plant growth hormone belonging to the auxin class. It is utilized in various biological applications.
  • Acts as a natural plant growth hormone.
  • Induces plant cell elongation and division in cell culture mediums.
  • Functions as a tracer in research.
  • Serves as an internal standard for quantitative analysis.
  • Compatible with analytical techniques such as NMR, GC-MS, and LC-MS.