Learn More
Medchemexpress LLC 3-Indoleacetic acid | 87-51-4 | 99.9% | 175.19 | 10 G
SDP
Catalog No. 5000387966
Encompass_Preferred
3-Indoleacetic acid is an IAA hormone and growth regulator that promotes plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. It is for research use only.
  • An IAA hormone and growth regulator.
  • Promotes plant nutritional growth through cell expansion, differentiation, morphogenesis, and organogenesis.
  • Promotes the enrichment of salt tolerant bacteria with growth promoting function during the secondary fermentation process of compost.
  • Improves outcrossing rate of diverse rice cytoplasmic male sterile lines and hybrid rice seed production through affecting floral traits (at 50 g/ha).
  • Improves photosynthetic performance and reduces the toxic effect of cadmium on C. camphora (at 10 mg/kg).
Catalog No. 50-003-87966 Supplier Medchemexpress LLC Supplier No. HY18569B10G
3-Indoleacetic acid is an IAA hormone and growth regulator that promotes plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. It is for research use only.
  • An IAA hormone and growth regulator.
  • Promotes plant nutritional growth through cell expansion, differentiation, morphogenesis, and organogenesis.
  • Promotes the enrichment of salt tolerant bacteria with growth promoting function during the secondary fermentation process of compost.
  • Improves outcrossing rate of diverse rice cytoplasmic male sterile lines and hybrid rice seed production through affecting floral traits (at 50 g/ha).
  • Improves photosynthetic performance and reduces the toxic effect of cadmium on C. camphora (at 10 mg/kg).