Learn More
Medchemexpress LLC 3-Indoleacetic acid | 87-51-4 | 99.9% | 175.19 | 25 G
SDP
Catalog No. 5000385571
Encompass_Preferred
3-Indoleacetic acid is an IAA hormone and growth regulator that promotes plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. It is intended for research use only.
  • Iaa hormone and growth regulator.
  • Can promote plant nutritional growth.
  • Involved in cell expansion, differentiation, morphogenesis, and organogenesis.
  • Purity of 99.94%.
  • Available in various solid and solution quantities.
  • Related to numerous publications citing its use in research.
Catalog No. 50-003-85571 Supplier Medchemexpress LLC Supplier No. 25GHY18569B
3-Indoleacetic acid is an IAA hormone and growth regulator that promotes plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. It is intended for research use only.
  • Iaa hormone and growth regulator.
  • Can promote plant nutritional growth.
  • Involved in cell expansion, differentiation, morphogenesis, and organogenesis.
  • Purity of 99.94%.
  • Available in various solid and solution quantities.
  • Related to numerous publications citing its use in research.