Learn More
Medchemexpress LLC 3-Indoleacetic acid | 87-51-4 | 99.9% | 175.19 | 25 MG
SDP
Catalog No. 5000384110
Encompass_Preferred
3-Indoleacetic acid is an IAA hormone and growth regulator that promotes plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. This product is for research use only and is not intended for patient use.
  • Promotes the enrichment of salt tolerant bacteria with growth promoting function during the secondary fermentation process of compost.
  • Improves outcrossing rate of diverse rice cytoplasmic male lines and hybrid rice seed production.
  • Improves photosynthetic performance and reduces the toxic effect of cadmium on *C. camphora*.
Catalog No. 50-003-84110 Supplier Medchemexpress LLC Supplier No. 25MGHY18569R
3-Indoleacetic acid is an IAA hormone and growth regulator that promotes plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. This product is for research use only and is not intended for patient use.
  • Promotes the enrichment of salt tolerant bacteria with growth promoting function during the secondary fermentation process of compost.
  • Improves outcrossing rate of diverse rice cytoplasmic male lines and hybrid rice seed production.
  • Improves photosynthetic performance and reduces the toxic effect of cadmium on *C. camphora*.