Learn More
Medchemexpress LLC 3-Indoleacetic acid | 87-51-4 | 99.9% | 175.19 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY18569R
Encompass_Preferred
3-Indoleacetic acid is an IAA hormone and growth regulator that promotes plant nutritional growth. It is involved in various crucial plant processes including cell expansion, differentiation, morphogenesis, and organogenesis. This product is intended for research purposes only and is not suitable for patient use. It typically presents as an off-white to light brown solid.
  • Promotes plant cell expansion
  • Supports plant cell differentiation
  • Aids in plant morphogenesis
  • Contributes to plant organogenesis
  • Functions as a plant growth regulator
Catalog No. 50-003-85544