Learn More
Medchemexpress LLC 3-Indoleacetic acid | 87-51-4 | MFCD00005636 | 99.8% | 175.19 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY18569R100MG
Encompass_Preferred
3-Indoleacetic acid (Standard) is the analytical standard of 3-Indoleacetic acid. This product is intended for research and analytical applications. 3-Indoleacetic acid is an IAA hormone and growth regulator that can promote plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-87829