Learn More
Medchemexpress LLC 3-Indoleacetic acid | 87-51-4 | MFCD00005707 | ≥98% | 175.18 | 1 MG
SDP
Catalog No. 5000387938
Encompass_Preferred
3-Indoleacetic acid (IAA) is the most common natural plant growth hormone of the auxin class. It can promote plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. It can also be added to cell culture medium to induce plant cell elongation and division. Additionally, 3-Indoleacetic acid protects bacteria from toxic damage to cell membrane potential caused by other indole intermediate products.
  • Promotes plant nutritional growth through cell expansion, differentiation, morphogenesis, and organogenesis.
  • Induces plant cell elongation and division when added to cell culture medium.
  • Protects bacteria from toxic damage to cell membrane potential.
  • Functions as an IAA hormone and growth regulator.
Catalog No. 50-003-87938 Supplier Medchemexpress LLC Supplier No. HY18569S31MG
3-Indoleacetic acid (IAA) is the most common natural plant growth hormone of the auxin class. It can promote plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. It can also be added to cell culture medium to induce plant cell elongation and division. Additionally, 3-Indoleacetic acid protects bacteria from toxic damage to cell membrane potential caused by other indole intermediate products.
  • Promotes plant nutritional growth through cell expansion, differentiation, morphogenesis, and organogenesis.
  • Induces plant cell elongation and division when added to cell culture medium.
  • Protects bacteria from toxic damage to cell membrane potential.
  • Functions as an IAA hormone and growth regulator.