Learn More
Medchemexpress LLC 3-Indoleacetic acid-d4 | 76937-77-4 | 99.4% | 179.21 | 2.5 MG
SDP
Catalog No. 5000387839
Encompass_Preferred
3-Indoleacetic acid-d4 (Indole-3-acetic acid-d4; 3-IAA-d4) is the deuterium labeled 3-Indoleacetic acid, which is the most common natural plant growth hormone of the auxin class. It can be used to induce plant cell elongation and division in cell culture medium.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-87839 Supplier Medchemexpress LLC Supplier No. HY18569S12.5MG
3-Indoleacetic acid-d4 (Indole-3-acetic acid-d4; 3-IAA-d4) is the deuterium labeled 3-Indoleacetic acid, which is the most common natural plant growth hormone of the auxin class. It can be used to induce plant cell elongation and division in cell culture medium.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS