Learn More
Medchemexpress LLC 3-Indoleacetic acid-d5 | 76937-78-5 | 99.76% | 180.21 | 1 ML
SDP
Catalog No. 5000387830
Encompass_Preferred
The product, 3-Indoleacetic acid-d5 (also known as Indole-3-acetic acid-d5 or 3-IAA-d5), is a deuterium-labeled variant of 3-Indoleacetic acid. This compound is utilized as an internal standard for analyzing IAA releases that occur after the alkaline hydrolysis of ester and amide conjugates. It also serves as a tracer in various applications.
  • It is a stable isotope-labeled compound.
  • Used for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeling can influence pharmacokinetic and metabolic profiles of drugs.
  • High purity of 99.76%.
Catalog No. 50-003-87830 Supplier Medchemexpress LLC Supplier No. HY18569S10MM/1ML
The product, 3-Indoleacetic acid-d5 (also known as Indole-3-acetic acid-d5 or 3-IAA-d5), is a deuterium-labeled variant of 3-Indoleacetic acid. This compound is utilized as an internal standard for analyzing IAA releases that occur after the alkaline hydrolysis of ester and amide conjugates. It also serves as a tracer in various applications.
  • It is a stable isotope-labeled compound.
  • Used for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeling can influence pharmacokinetic and metabolic profiles of drugs.
  • High purity of 99.76%.