Learn More
Medchemexpress LLC 3-Indoleacetic acid-d5 | 76937-78-5 | 99.8% | 180.21 | 1 MG
SDP
Catalog No. 5000387923
Encompass_Preferred
3-Indoleacetic acid-d5 is the deuterium labeled 3-Indoleacetic acid (HY-18569). It can be used as an internal standard for assay of IAA releases by alkaline hydrolysis of ester and amide conjugates. It is for research use only and not sold to patients.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-87923 Supplier Medchemexpress LLC Supplier No. HY18569S1MG
3-Indoleacetic acid-d5 is the deuterium labeled 3-Indoleacetic acid (HY-18569). It can be used as an internal standard for assay of IAA releases by alkaline hydrolysis of ester and amide conjugates. It is for research use only and not sold to patients.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.