Learn More
Medchemexpress LLC 3-Indoleacetic acid-d7 | 1173020-21-7 | 99.8% | 182.23 | 5 MG
SDP
Catalog No. 5000387939
Encompass_Preferred
3-Indoleacetic acid-d7 is a deuterium-labeled derivative of 3-Indoleacetic acid (HY-18569), the most common natural plant growth hormone of the auxin class. It can be utilized in cell culture mediums to promote plant cell elongation and division. This compound serves as a valuable tool for research applications, including its use as a tracer and an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Induces plant cell elongation and division
Catalog No. 50-003-87939 Supplier Medchemexpress LLC Supplier No. HY18569S35MG
3-Indoleacetic acid-d7 is a deuterium-labeled derivative of 3-Indoleacetic acid (HY-18569), the most common natural plant growth hormone of the auxin class. It can be utilized in cell culture mediums to promote plant cell elongation and division. This compound serves as a valuable tool for research applications, including its use as a tracer and an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Induces plant cell elongation and division