Learn More
Medchemexpress LLC 3-Indoleacetic acid (sodium) | 6505-45-9 | 100.0% | 197.17 | 500 MG
SDP
Catalog No. 5000387820
Encompass_Preferred
3-Indoleacetic acid sodium is an IAA hormone and growth regulator that can promote plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. It is for research use only and not sold to patients.
  • IAA hormone and growth regulator.
  • Promotes plant nutritional growth through cell expansion, differentiation, morphogenesis, and organogenesis.
  • Purity: 99.95%.
  • Available in solid and solution forms.
  • Promotes enrichment of salt tolerant bacteria.
  • Improves outcrossing rate in rice.
  • Reduces toxic effect of cadmium on C. camphora.
  • Significantly inhibited tumor growth in dose-dependent manner.
Catalog No. 50-003-87820 Supplier Medchemexpress LLC Supplier No. HY18569A500MG
3-Indoleacetic acid sodium is an IAA hormone and growth regulator that can promote plant nutritional growth through processes such as cell expansion, differentiation, morphogenesis, and organogenesis. It is for research use only and not sold to patients.
  • IAA hormone and growth regulator.
  • Promotes plant nutritional growth through cell expansion, differentiation, morphogenesis, and organogenesis.
  • Purity: 99.95%.
  • Available in solid and solution forms.
  • Promotes enrichment of salt tolerant bacteria.
  • Improves outcrossing rate in rice.
  • Reduces toxic effect of cadmium on C. camphora.
  • Significantly inhibited tumor growth in dose-dependent manner.