Learn More
Medchemexpress LLC 3-Indoleacetic acid sodium | 6505-45-9 | 99.95% | 197.17 | 100 MG
SDP
Catalog No. 5000387964
Encompass_Preferred
3-Indoleacetic acid sodium is an IAA hormone and growth regulator that promotes plant nutritional growth through processes like cell expansion, differentiation, morphogenesis, and organogenesis. This product is for research use only and not sold to patients. Its benefits include enhancing composting processes, improving rice outcrossing rates, and mitigating cadmium toxicity in certain plants.
  • Functions as an IAA hormone and growth regulator
  • Promotes plant nutritional growth
  • Promotes enrichment of salt tolerant bacteria with growth promoting function during secondary fermentation of compost
  • Improves outcrossing rate of diverse rice cytoplasmic male sterile lines and hybrid rice seed production through affecting floral traits
  • Improves photosynthetic performance and reduces the toxic effect of cadmium on C. camphora
Catalog No. 50-003-87964 Supplier Medchemexpress LLC Supplier No. HY18569A100MG
3-Indoleacetic acid sodium is an IAA hormone and growth regulator that promotes plant nutritional growth through processes like cell expansion, differentiation, morphogenesis, and organogenesis. This product is for research use only and not sold to patients. Its benefits include enhancing composting processes, improving rice outcrossing rates, and mitigating cadmium toxicity in certain plants.
  • Functions as an IAA hormone and growth regulator
  • Promotes plant nutritional growth
  • Promotes enrichment of salt tolerant bacteria with growth promoting function during secondary fermentation of compost
  • Improves outcrossing rate of diverse rice cytoplasmic male sterile lines and hybrid rice seed production through affecting floral traits
  • Improves photosynthetic performance and reduces the toxic effect of cadmium on C. camphora