Learn More
Medchemexpress LLC 3-indoleacrylic acid | 1204-06-4 | MFCD00005633 | 99.4% | 187.20 g·mol⁻¹ | C11H9NO2 | 1 ML
SDP
Catalog No. 5000303697
Encompass_Preferred
3-Indoleacrylic acid (CAS 1204-06-4) is an indole-derived bioactive compound supplied as a 10 mM solution in DMSO for research use. It exhibits antialgal activity by increasing reactive oxygen species (ROS), lowering mitochondrial membrane potential, disrupting antioxidant systems, and down-regulating photosynthetic and nutrient-assimilation genes in target organisms. The product is provided at high purity for laboratory studies of ROS, mitochondrial function, and gene expression.
Catalog No. 50-003-03697 Supplier Medchemexpress LLC Supplier No. HYW01527310MM/1ML
3-Indoleacrylic acid (CAS 1204-06-4) is an indole-derived bioactive compound supplied as a 10 mM solution in DMSO for research use. It exhibits antialgal activity by increasing reactive oxygen species (ROS), lowering mitochondrial membrane potential, disrupting antioxidant systems, and down-regulating photosynthetic and nutrient-assimilation genes in target organisms. The product is provided at high purity for laboratory studies of ROS, mitochondrial function, and gene expression.