Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-indoleacrylic acid-d4 | 99.4% | 191.22 g/mol | C11H5D4NO2 | 5 MG
SDP

Catalog No. 5000302564
Encompass_Preferred

3-Indoleacrylic acid-d4 is a deuterium-labeled analogue of 3-indoleacrylic acid supplied as a research reagent and stable isotope standard for analytical applications. The deuterated tracer provides a mass-shifted reference while preserving the parent compound's chemical behavior, supporting quantitation and metabolic studies.

  • Deuterium-labeled (d4) for use as an internal standard in mass spectrometry and isotopic tracing.
  • High purity as supplied (99.38% HPLC).
  • Molecular weight 191.22 g/mol and formula C11H5D4NO2.
  • Available in small analytical sizes (1-50 mg).
  • Store powder at -20°C; in solvent store at -80°C for long-term stability.

Catalog No. 50-003-02564 Supplier Medchemexpress LLC Supplier No. HYW015273S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Indoleacrylic acid-d4 is a deuterium-labeled analogue of 3-indoleacrylic acid supplied as a research reagent and stable isotope standard for analytical applications. The deuterated tracer provides a mass-shifted reference while preserving the parent compound's chemical behavior, supporting quantitation and metabolic studies.

  • Deuterium-labeled (d4) for use as an internal standard in mass spectrometry and isotopic tracing.
  • High purity as supplied (99.38% HPLC).
  • Molecular weight 191.22 g/mol and formula C11H5D4NO2.
  • Available in small analytical sizes (1-50 mg).
  • Store powder at -20°C; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.