Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Indoleacrylic Acid 98.0+%, TCI America™
SDP

Catalog No. I00251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00251G 1 g
I00255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00251G Supplier TCI America Supplier No. I00251G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1204-06-4
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.198
MDL Number MFCD00005633
InChI Key PLVPPLCLBIEYEA-AATRIKPKSA-N
Synonym 3-indoleacrylic acid, trans-3-indoleacrylic acid, indole-3-acrylic acid, 3-indolylacrylic acid, indole-3beta-acrylic acid, 3-1h-indol-3-yl acrylic acid, indole-3-crylic acid, 2e-3-1h-indol-3-yl prop-2-enoic acid, trans-3-1h-indol-3-yl acrylic acid, indoleacrylic acid van
PubChem CID 5375048
IUPAC Name (E)-3-(1H-indol-3-yl)prop-2-enoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)C=CC(=O)O

Specifications

Quantity 1 g
Formula Weight 187.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indoleacrylic Acid
RTECSNumber NL3680000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.