Learn More
Apexbio Technology LLC 3-Indolebutyric acid (IBA) 133-32-4 50g
SDP
Catalog No. 5000090246
Encompass_Preferred
3-Indolebutyric acid (IBA CAS 133-32-4) is a synthetic auxin commonly employed in plant biology for its role in promoting root and shoot formation in tissue culture systems Mechanistically IBA acts as a plant growth regulator facilitating cell division and differentiation Beyond plant research IBA has been investigated in animal models notably for enhancing embryonic development in avian eggs Application of optimal IBA concentrations to fertilized poultry eggs has been shown to increase the proportion of viable embryos and improve overall hatch rates even in cases where initial quality assessment did not indicate embryonic viability Thus IBA is relevant for studies of developmental biology and agricultural biotechnology
Catalog No. 50-000-90246 Supplier Apexbio Technology LLC Supplier No. B598550000
3-Indolebutyric acid (IBA CAS 133-32-4) is a synthetic auxin commonly employed in plant biology for its role in promoting root and shoot formation in tissue culture systems Mechanistically IBA acts as a plant growth regulator facilitating cell division and differentiation Beyond plant research IBA has been investigated in animal models notably for enhancing embryonic development in avian eggs Application of optimal IBA concentrations to fertilized poultry eggs has been shown to increase the proportion of viable embryos and improve overall hatch rates even in cases where initial quality assessment did not indicate embryonic viability Thus IBA is relevant for studies of developmental biology and agricultural biotechnology