Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Indolebutyric Acid 98.0+%, TCI America™
SDP

Catalog No. I002625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I002625G 25 g
I00265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I002625G Supplier TCI America Supplier No. I002625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133-32-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00005664
InChI Key JTEDVYBZBROSJT-UHFFFAOYSA-N
Synonym indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
PubChem CID 8617
ChEBI CHEBI:33070
IUPAC Name 4-(1H-indol-3-yl)butanoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 203.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indolebutyric Acid
EINECSNumber (5)-3368&(9)-0297
RTECSNumber NL5250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.