Learn More
Medchemexpress LLC 3-Indolepropionic acid | 830-96-6 | MFCD00005660 | >98.0% | 189.21 g/mol | C11H11NO2 | 100 MG
SDP
Catalog No. 5000303209
Encompass_Preferred
3-Indolepropionic acid (indole-3-propionic acid, 3-IPA) is an endogenous metabolite supplied as an analytical standard for research and analytical applications. It is an off-white powder with molecular formula C11H11NO2, molecular weight 189.21 g/mol, and CAS 830-96-6. The compound is commonly used as a reference in antioxidant, metabolic, and biochemical assays.
Catalog No. 50-003-03209 Supplier Medchemexpress LLC Supplier No. HYW015229R100MG
3-Indolepropionic acid (indole-3-propionic acid, 3-IPA) is an endogenous metabolite supplied as an analytical standard for research and analytical applications. It is an off-white powder with molecular formula C11H11NO2, molecular weight 189.21 g/mol, and CAS 830-96-6. The compound is commonly used as a reference in antioxidant, metabolic, and biochemical assays.