Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Indolepropionic acid | 830-96-6 | MFCD00005660 | 99.9% | 189.21 g·mol⁻1 | C11H11NO2 | 10 G
SDP

Catalog No. 5000302560
Encompass_Preferred

3-Indolepropionic acid is an endogenous indole-derived metabolite and a potent antioxidant widely used in research on neuroprotection and Alzheimer's disease. Supplied as an off-white to yellow powder, it is suitable for in vitro and in vivo studies and includes manufacturer guidance for solubility and storage.

  • Potent antioxidant and neuroprotective agent.
  • Endogenous metabolite relevant to biochemical and pharmacological studies.
  • High analytical purity suitable for research applications.
  • Off-white to yellow powder for convenient handling.
  • Soluble in DMSO (≥100 mg/mL); water solubility improves with warming and sonication.
  • Storage: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years); solutions require colder storage.
  • Manufacturer provides in vivo vehicle formulations for dosing.

Catalog No. 50-003-02560 Supplier Medchemexpress LLC Supplier No. HYW01522910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Indolepropionic acid is an endogenous indole-derived metabolite and a potent antioxidant widely used in research on neuroprotection and Alzheimer's disease. Supplied as an off-white to yellow powder, it is suitable for in vitro and in vivo studies and includes manufacturer guidance for solubility and storage.

  • Potent antioxidant and neuroprotective agent.
  • Endogenous metabolite relevant to biochemical and pharmacological studies.
  • High analytical purity suitable for research applications.
  • Off-white to yellow powder for convenient handling.
  • Soluble in DMSO (≥100 mg/mL); water solubility improves with warming and sonication.
  • Storage: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years); solutions require colder storage.
  • Manufacturer provides in vivo vehicle formulations for dosing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.