Learn More
Medchemexpress LLC 3-Indolyl acetate | 608-08-2 | 99.26% | 175.18 | 100 MG
SDP
Catalog No. 5000352219
Encompass_Preferred
3-Indolyl acetate is an ester derivative of indole, found in various plant tissues, particularly in cruciferous vegetables. It acts as an important signaling molecule in plants, regulating physiological processes such as growth and defense against pathogens. It also shows potential in the pharmaceutical industry due to observed antitumor and anti-inflammatory effects in some studies. As a biochemical reagent, it can be used for life science research.
  • Ester derivative of indole
  • Found in various plant tissues
  • Regulates physiological processes in plants
  • Potential antitumor effects
  • Potential anti-inflammatory effects
  • Used as a biochemical reagent for life science research
Catalog No. 50-003-52219 Supplier Medchemexpress LLC Supplier No. HY137884100MG
3-Indolyl acetate is an ester derivative of indole, found in various plant tissues, particularly in cruciferous vegetables. It acts as an important signaling molecule in plants, regulating physiological processes such as growth and defense against pathogens. It also shows potential in the pharmaceutical industry due to observed antitumor and anti-inflammatory effects in some studies. As a biochemical reagent, it can be used for life science research.
  • Ester derivative of indole
  • Found in various plant tissues
  • Regulates physiological processes in plants
  • Potential antitumor effects
  • Potential anti-inflammatory effects
  • Used as a biochemical reagent for life science research