Learn More
Medchemexpress LLC 3-Indolyl acetate | 608-08-2 | 99.3% | 175.18 | 1 G
SDP
Catalog No. 5000351944
Encompass_Preferred
3-Indolyl acetate is an ester derivative of indole, widely present in various plant tissues, particularly cruciferous vegetables like broccoli and cabbage. This biochemical reagent functions as an important signaling molecule in plants, regulating physiological processes such as growth and defense against pathogens. It has also shown potential applications in the pharmaceutical industry due to its demonstrated antitumor and anti-inflammatory effects in various studies.
  • Ester derivative of indole
  • Found in various plant tissues, including cruciferous vegetables
  • Acts as an important signaling molecule in plants
  • Regulates plant growth and defense against pathogens
  • Potential pharmaceutical applications
  • Demonstrates antitumor effects in some studies
  • Demonstrates anti-inflammatory effects in some studies
  • Suitable for life science research as a biological material or organic compound
Catalog No. 50-003-51944 Supplier Medchemexpress LLC Supplier No. HY1378841G
3-Indolyl acetate is an ester derivative of indole, widely present in various plant tissues, particularly cruciferous vegetables like broccoli and cabbage. This biochemical reagent functions as an important signaling molecule in plants, regulating physiological processes such as growth and defense against pathogens. It has also shown potential applications in the pharmaceutical industry due to its demonstrated antitumor and anti-inflammatory effects in various studies.
  • Ester derivative of indole
  • Found in various plant tissues, including cruciferous vegetables
  • Acts as an important signaling molecule in plants
  • Regulates plant growth and defense against pathogens
  • Potential pharmaceutical applications
  • Demonstrates antitumor effects in some studies
  • Demonstrates anti-inflammatory effects in some studies
  • Suitable for life science research as a biological material or organic compound