Learn More
Medchemexpress LLC 3-Indolyl acetate | 608-08-2 | 99.3% | 175.18 | 250 MG
SDP
Catalog No. 5000349392
Encompass_Preferred
3-Indolyl acetate is an ester derivative of indole found in various plant tissues, particularly in cruciferous vegetables. It acts as an important signaling molecule in plants, regulating physiological processes such as growth and defense against pathogens. Additionally, it shows potential in the pharmaceutical industry due to demonstrated antitumor and anti-inflammatory effects, making it suitable as a biochemical reagent for life science-related research.
  • Ester derivative of indole
  • Found in various plant tissues
  • Regulates plant growth and defense mechanisms
  • Exhibits potential antitumor effects
  • Displays anti-inflammatory properties
  • Suitable as a biochemical reagent for life science research
Catalog No. 50-003-49392 Supplier Medchemexpress LLC Supplier No. 250MGHY137884
3-Indolyl acetate is an ester derivative of indole found in various plant tissues, particularly in cruciferous vegetables. It acts as an important signaling molecule in plants, regulating physiological processes such as growth and defense against pathogens. Additionally, it shows potential in the pharmaceutical industry due to demonstrated antitumor and anti-inflammatory effects, making it suitable as a biochemical reagent for life science-related research.
  • Ester derivative of indole
  • Found in various plant tissues
  • Regulates plant growth and defense mechanisms
  • Exhibits potential antitumor effects
  • Displays anti-inflammatory properties
  • Suitable as a biochemical reagent for life science research