Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Indoxyl acetate, 97%

Catalog No. AAL0493203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0493203 1 g
AAL0493206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0493203 Supplier Thermo Scientific Chemicals Supplier No. L0493203
Add to Cart

CAS: 608-08-2 | C10H9NO2 | 175.187 g/mol

Useful for detection of esterases3-Indoxyl acetate acts as a colorimetric substrate used for differentiating esterases from various microbiological species. Further, it is used as a plant growth-stimulating hormone and reagent. In addition to this, it acts as a useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful for detection of esterases3-Indoxyl acetate acts as a colorimetric substrate used for differentiating esterases from various microbiological species. Further, it is used as a plant growth-stimulating hormone and reagent. In addition to this, it acts as a useful synthetic intermediate.

Solubility
Soluble in ethanol, water, chloroform, dichloromethane and methanol.

Notes
Light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-08-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00014561
InChI Key JBOPQACSHPPKEP-UHFFFAOYSA-N
Synonym 3-acetoxyindole, indoxyl acetate, 3-indoxyl acetate, indoxyl-o-acetate, 3-indolyl acetate, indole, 3-acetato, indoxylacetate, 1h-indol-3-ol, 3-acetate, 1h-indol-3-ol, acetate ester, acetic acid, 3-indolyl ester
PubChem CID 11841
IUPAC Name 1H-indol-3-yl acetate
SMILES CC(=O)OC1=CNC2=CC=CC=C21

Specifications

Melting Point 128°C to 130°C
Quantity 1 g
Beilstein 143086
Solubility Information Soluble in ethanol,water,chloroform,dichloromethane and methanol.
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 3-Indoxyl acetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-154-2
RTECSNumber AI3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.